(3-Methoxy-2,4-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49755-500MG 49755-1G 49755-5G
77338-248EA 470.71 USD
77338-248 77351-596 77375-870
(3-Methoxy-2,4-dimethylphenyl)(methyl)sulfane ≥97%
(3-Methoxy-2,4-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28795058
CAS Number: 1806334-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR