1-Iodo-3-methoxy-2,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

49754-500MG 49754-1G 49754-5G
77336-036EA 374.92 USD
77336-036 77351-594 77375-868
1-Iodo-3-methoxy-2,4-dimethylbenzene ≥97%
1-Iodo-3-methoxy-2,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
MW: 262.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794895
CAS Number: 135630-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR