Ethyl 3-methoxy-2,4-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49751-500MG 49751-1G 49751-5G
77339-266EA 571.65 USD
77339-266 77351-592 77375-866
Ethyl 3-methoxy-2,4-dimethylbenzoate ≥97%
Ethyl 3-methoxy-2,4-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD28794950
CAS Number: 1806570-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR