Methyl 3-methoxy-2,4-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49750-500MG 49750-1G 49750-5G
77339-264EA 571.65 USD
77339-264 77351-590 77375-864
Methyl 3-methoxy-2,4-dimethylbenzoate ≥97%
Methyl 3-methoxy-2,4-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD28794957
CAS Number: 1803854-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR