2,5-Dichloro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49736-500MG 49736-1G 49736-5G 49736-25G
77328-052EA 256.47 USD
77328-052 77351-586 77364-382 77386-690
2,5-Dichloro-N,N-dimethylbenzamide ≥97%
2,5-Dichloro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD01195594
CAS Number: 90526-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR