2,5-Dichloro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49735-1G 49735-5G 49735-25G
77341-766EA 104.03 USD
77341-766 77364-380 77375-860
2,5-Dichloro-N-methylbenzamide ≥97%
2,5-Dichloro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD01195593
CAS Number: 5980-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR