(2,5-Dichlorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49733-1G 49733-5G 49733-25G
77340-696EA 60.77 USD
77340-696 77364-376 77375-856
(2,5-Dichlorophenyl)(thiomorpholino)methanone ≥97%
(2,5-Dichlorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁Cl₂NOS
MW: 276.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756626
CAS Number: 1626053-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR