2,5-Dichloro-N-cyclohexylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49729-500MG 49729-1G 49729-5G 49729-25G
77328-050EA 242.05 USD
77328-050 77351-584 77364-370 77386-688
2,5-Dichloro-N-cyclohexylbenzamide ≥97%
2,5-Dichloro-N-cyclohexylbenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD01195571
CAS Number: 329930-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR