2,5-Dichloro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49725-500MG 49725-1G 49725-5G
77339-262EA 571.65 USD
77339-262 77351-582 77375-848
2,5-Dichloro-3-formylbenzoic acid ≥97%
2,5-Dichloro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄Cl₂O₃
MW: 219.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564149
CAS Number: 2384099-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR