2-Fluoro-5-iodo-1-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

49724-500MG 49724-1G 49724-5G 49724-25G
77328-048EA 214.24 USD
77328-048 77351-580 77364-366 77386-686
2-Fluoro-5-iodo-1-methoxy-3-methylbenzene ≥97%
2-Fluoro-5-iodo-1-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD34530016
CAS Number: 2385912-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR