(2-Fluoro-5-iodo-3-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49723-500MG 49723-1G 49723-5G 49723-25G
77328-046EA 136.99 USD
77328-046 77344-532 77364-364 77386-684
(2-Fluoro-5-iodo-3-methoxyphenyl)(methyl)sulfane ≥97%
(2-Fluoro-5-iodo-3-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIOS
MW: 298.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564148
CAS Number: 2711470-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR