1-Chloro-2-fluoro-5-iodo-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49722-500MG 49722-1G 49722-5G 49722-25G
77328-044EA 236.9 USD
77328-044 77351-578 77364-362 77386-682
1-Chloro-2-fluoro-5-iodo-3-methoxybenzene ≥97%
1-Chloro-2-fluoro-5-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756625
CAS Number: 2386206-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR