Ethyl 2-fluoro-5-iodo-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

49719-500MG 49719-1G 49719-5G 49719-25G
77328-040EA 220.42 USD
77328-040 77351-576 77364-358 77386-678
Ethyl 2-fluoro-5-iodo-3-methoxybenzoate ≥97%
Ethyl 2-fluoro-5-iodo-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀FIO₃
MW: 324.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756624
CAS Number: 2387111-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR