Methyl 2-fluoro-5-iodo-3-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

49718-500MG 49718-1G 49718-5G 49718-25G
77328-038EA 222.48 USD
77328-038 77351-574 77364-356 77386-676
Methyl 2-fluoro-5-iodo-3-methoxybenzoate ≥95%
Methyl 2-fluoro-5-iodo-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈FIO₃
MW: 310.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756623
CAS Number: 2092791-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR