2-Fluoro-5-iodo-3-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
49717-500MG
49717-1G
49717-5G
49717-25G
77328-036EA
187.46
USD
77328-036
77351-572
77364-354
77386-674
2-Fluoro-5-iodo-3-methoxybenzoic acid ≥97%
2-Fluoro-5-iodo-3-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆FIO₃ Storage Temperature: Ambient |
MDL Number:
MFCD34756622 CAS Number: 2090422-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...