(2-Fluoro-5-iodo-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

49716-500MG 49716-1G 49716-5G 49716-25G
77328-034EA 225.57 USD
77328-034 77351-570 77364-352 77386-672
(2-Fluoro-5-iodo-3-methoxyphenyl)methanol ≥97%
(2-Fluoro-5-iodo-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈FIO₂
MW: 282.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756621
CAS Number: 2091048-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR