2-Fluoro-5-iodo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49715-500MG 49715-1G 49715-5G 49715-25G
77328-032EA 187.46 USD
77328-032 77351-568 77364-350 77386-670
2-Fluoro-5-iodo-3-methoxybenzaldehyde ≥97%
2-Fluoro-5-iodo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34564145
CAS Number: 2092471-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR