(2-Bromo-5-chlorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49700-500MG 49700-1G 49700-5G 49700-25G
77328-028EA 105.06 USD
77328-028 77343-074 77364-346 77375-842
(2-Bromo-5-chlorophenyl)(methyl)sulfane ≥97%
(2-Bromo-5-chlorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₆BrClS
Storage Temperature: Ambient
MDL Number: MFCD22488881
CAS Number: 255051-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR