(4-Chloro-2-ethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

49699-500MG 49699-1G 49699-5G
77328-026EA 338.87 USD
77328-026 77351-564 77375-840
(4-Chloro-2-ethoxyphenyl)methanol ≥97%
(4-Chloro-2-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD27930720
CAS Number: 869088-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR