5-Chloro-3-fluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49693-500MG 49693-1G 49693-5G 49693-25G
77328-022EA 187.46 USD
77328-022 77351-562 77364-342 77386-666
5-Chloro-3-fluoro-2-methoxybenzoic acid ≥97%
5-Chloro-3-fluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
MW: 204.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415164
CAS Number: 1545525-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR