(5-Chloro-3-fluoro-2-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

49692-500MG 49692-1G 49692-5G 49692-25G
77328-020EA 280.16 USD
77328-020 77351-560 77364-340 77386-664
(5-Chloro-3-fluoro-2-methoxyphenyl)methanol ≥97%
(5-Chloro-3-fluoro-2-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28129275
CAS Number: 1444384-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR