1-Bromo-3-iodo-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

49691-500MG 49691-1G 49691-5G
77336-732EA 393.46 USD
77336-732 77351-558 77375-838
1-Bromo-3-iodo-2,5-dimethylbenzene ≥97%
1-Bromo-3-iodo-2,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrI
MW: 310.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738491
CAS Number: 1807198-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR