1-Bromo-3-chloro-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

49688-500MG 49688-1G 49688-5G 49688-25G
77328-018EA 201.88 USD
77328-018 77351-556 77364-338 77386-662
1-Bromo-3-chloro-2,5-dimethylbenzene ≥97%
1-Bromo-3-chloro-2,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MW: 219.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178575
CAS Number: 1780808-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR