Ethyl 3-bromo-2,5-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

49686-500MG 49686-1G 49686-5G
77337-370EA 412 USD
77337-370 77351-554 77375-836
Ethyl 3-bromo-2,5-dimethylbenzoate ≥95%
Ethyl 3-bromo-2,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738473
CAS Number: 1805026-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR