1-Chloro-3-ethoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

49678-500MG 49678-1G 49678-5G 49678-25G
77328-016EA 197.76 USD
77328-016 77351-552 77364-334 77386-660
1-Chloro-3-ethoxy-5-(trifluoromethyl)benzene ≥97%
1-Chloro-3-ethoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈ClF₃O
MW: 224.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044198
CAS Number: 1881332-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR