1-(Benzyloxy)-3-chloro-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

49677-500MG 49677-1G 49677-25G 49677-5G
77328-014EA 299.73 USD
77328-014 77351-550 77386-658 77375-832
1-(Benzyloxy)-3-chloro-5-(trifluoromethyl)benzene ≥97%
1-(Benzyloxy)-3-chloro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClF₃O
MW: 286.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902846
CAS Number: 2586125-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR