Ethyl 1-(tert-butyl)-1H-pyrazole-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

49672-500MG 49672-1G 49672-5G
77338-072EA 462.47 USD
77338-072 77351-548 77375-830
Ethyl 1-(tert-butyl)-1H-pyrazole-5-carboxylate ≥97%
Ethyl 1-(tert-butyl)-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21608590
CAS Number: 1416372-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR