1-(tert-Butyl)-5-iodo-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

49665-500MG 49665-1G 49665-5G 49665-25G
77328-012EA 126.69 USD
77328-012 77344-118 77364-332 77375-826
1-(tert-Butyl)-5-iodo-1H-pyrazole ≥97%
1-(tert-Butyl)-5-iodo-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁IN₂
Storage Temperature: Ambient
CAS Number: 1523609-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR