1-(tert-Butyl)-4-iodo-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

49662-500MG 49662-1G 49662-5G 49662-25G
77328-008EA 112.27 USD
77328-008 77343-302 77364-328 77375-822
1-(tert-Butyl)-4-iodo-1H-pyrazole ≥97%
1-(tert-Butyl)-4-iodo-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁IN₂
MW: 250.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22378621
CAS Number: 1354705-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR