Ethyl 4-ethoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49648-500MG 49648-1G 49648-5G 49648-25G
77328-004EA 128.75 USD
77328-004 77344-248 77364-326 77375-812
Ethyl 4-ethoxy-2-methylbenzoate ≥97%
Ethyl 4-ethoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31771931
CAS Number: 23676-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR