3-Fluoro-4-methoxy-2-methylphenol ≥95%

Supplier: AOBChem USA

Warning

49642-500MG 49642-1G 49642-5G
77339-010EA 544.87 USD
77339-010 77351-536 77375-808
3-Fluoro-4-methoxy-2-methylphenol ≥95%
3-Fluoro-4-methoxy-2-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD34530014
CAS Number: 1600530-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR