Methyl 3-fluoro-4-methoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49638-500MG 49638-1G 49638-5G
77338-332EA 477.92 USD
77338-332 77351-530 77375-802
Methyl 3-fluoro-4-methoxy-2-methylbenzoate ≥97%
Methyl 3-fluoro-4-methoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34181058
CAS Number: 2383417-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR