(3-Fluoro-4-methoxy-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49632-500MG 49632-1G 49632-5G
77339-214EA 568.56 USD
77339-214 77351-524 77375-796
(3-Fluoro-4-methoxy-2-methylphenyl)(methyl)sulfane ≥97%
(3-Fluoro-4-methoxy-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
MW: 186.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902844
CAS Number: 2586125-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR