5-Bromo-2-chloro-1-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

49630-500MG 49630-1G 49630-5G
77336-462EA 388.31 USD
77336-462 77351-522 77375-794
5-Bromo-2-chloro-1-iodo-3-methylbenzene ≥97%
5-Bromo-2-chloro-1-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClI
Storage Temperature: Ambient
MDL Number: MFCD34756611
CAS Number: 2385852-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR