Methyl 5-methyl-2-propoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

49621-500MG 49621-1G 49621-5G
77328-000EA 319.3 USD
77328-000 77351-518 77375-792
Methyl 5-methyl-2-propoxybenzoate ≥97%
Methyl 5-methyl-2-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD31775037
CAS Number: 1195353-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR