4-Bromo-2-chloro-5-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

49600-500MG 49600-1G 49600-5G 49600-25G
77327-996EA 263.68 USD
77327-996 77351-512 77364-318 77386-652
4-Bromo-2-chloro-5-fluorobenzotrifluoride ≥97%
4-Bromo-2-chloro-5-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂BrClF₄
Storage Temperature: Ambient
MDL Number: MFCD32869483
CAS Number: 2169005-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR