2,4-Dibromo-5-chloroaniline ≥97%

Supplier: AOBChem USA

Warning

49595-500MG 49595-1G 49595-5G 49595-25G
77327-994EA 241.02 USD
77327-994 77351-510 77364-316 77386-650
2,4-Dibromo-5-chloroaniline ≥97%
2,4-Dibromo-5-chloroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂ClN
Storage Temperature: Ambient
MDL Number: MFCD22480886
CAS Number: 89379-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR