1,3-Dibromo-2-chloro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

49575-500MG 49575-1G 49575-5G 49575-25G
77327-988EA 222.48 USD
77327-988 77351-508 77364-306 77386-646
1,3-Dibromo-2-chloro-5-iodobenzene ≥97%
1,3-Dibromo-2-chloro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClI
Storage Temperature: Ambient
MDL Number: MFCD34756597
CAS Number: 2387266-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR