6-(Hydroxymethyl)-2-naphthol ≥95%

Supplier: AOBChem USA

Warning

49473-1G 49473-5G 49473-25G
77340-690EA 60.77 USD
77340-690 77364-304 77375-778
6-(Hydroxymethyl)-2-naphthol ≥95%
6-(Hydroxymethyl)-2-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD12756488
CAS Number: 309752-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR