1-Fluoro-2-isopropoxy-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49451-500MG 49451-1G 49451-5G 49451-25G
77327-986EA 124.63 USD
77327-986 77344-054 77364-302 77375-776
1-Fluoro-2-isopropoxy-4-methoxybenzene ≥97%
1-Fluoro-2-isopropoxy-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
MW: 184.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092197
CAS Number: 1369855-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR