2-(Difluoromethoxy)-1-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49449-500MG 49449-1G 49449-5G
77337-652EA 434.66 USD
77337-652 77351-504 77375-772
2-(Difluoromethoxy)-1-fluoro-4-methoxybenzene ≥97%
2-(Difluoromethoxy)-1-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
MW: 192.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394919
CAS Number: 1261478-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR