1-(5-Ethoxy-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

49442-500MG 49442-1G 49442-5G
77327-980EA 322.39 USD
77327-980 77351-502 77375-768
1-(5-Ethoxy-2-fluorophenyl)ethanol ≥95%
1-(5-Ethoxy-2-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD32902836
CAS Number: 1554353-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR