(5-Bromo-2-fluoro-3-methoxyphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

49435-500MG 49435-1G 49435-5G 49435-25G
77327-974EA 233.81 USD
77327-974 77351-496 77364-292 77386-640
(5-Bromo-2-fluoro-3-methoxyphenyl)(piperidin-1-yl)methanone ≥97%
(5-Bromo-2-fluoro-3-methoxyphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrFNO₂
MW: 316.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403935
CAS Number: 2624417-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR