(5-Bromo-2-fluoro-3-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

49434-500MG 49434-1G 49434-5G 49434-25G
77327-972EA 214.24 USD
77327-972 77351-494 77364-290 77386-638
(5-Bromo-2-fluoro-3-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥95%
(5-Bromo-2-fluoro-3-methoxyphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrFNO₂
MW: 302.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902831
CAS Number: 2586126-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR