5-Bromo-2-fluoro-N,3-dimethoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49433-500MG 49433-1G 49433-5G 49433-25G
77327-970EA 189.52 USD
77327-970 77351-492 77364-288 77386-636
5-Bromo-2-fluoro-N,3-dimethoxy-N-methylbenzamide ≥97%
5-Bromo-2-fluoro-N,3-dimethoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrFNO₃
MW: 292.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902830
CAS Number: 2586127-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR