5-Bromo-N-cyclohexyl-2-fluoro-3-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

49432-500MG 49432-1G 49432-5G 49432-25G
77327-968EA 189.52 USD
77327-968 77351-490 77364-286 77386-634
5-Bromo-N-cyclohexyl-2-fluoro-3-methoxybenzamide ≥97%
5-Bromo-N-cyclohexyl-2-fluoro-3-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₇BrFNO₂
MW: 330.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902829
CAS Number: 2586126-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR