5-Bromo-2-fluoro-3-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49429-500MG 49429-1G 49429-5G 49429-25G
77327-962EA 189.52 USD
77327-962 77351-484 77364-280 77386-628
5-Bromo-2-fluoro-3-methoxy-N-methylbenzamide ≥97%
5-Bromo-2-fluoro-3-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD32902826
CAS Number: 2586126-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR