5-Bromo-2-fluoro-3-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

49428-500MG 49428-1G 49428-5G 49428-25G
77327-960EA 189.52 USD
77327-960 77351-482 77364-278 77386-626
5-Bromo-2-fluoro-3-methoxybenzamide ≥97%
5-Bromo-2-fluoro-3-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO₂
MW: 248.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902825
CAS Number: 2090556-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR