1,2-Dichloro-5-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

49421-500MG 49421-1G 49421-5G
77339-008EA 544.87 USD
77339-008 77351-474 77375-760
1,2-Dichloro-5-fluoro-3-iodobenzene ≥97%
1,2-Dichloro-5-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FI
MW: 290.889 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786113
CAS Number: 1806280-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR