Methyl 4-methoxy-2,3-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49420-500MG 49420-1G 49420-5G 49420-25G
77327-958EA 270.89 USD
77327-958 77351-472 77364-276 77386-624
Methyl 4-methoxy-2,3-dimethylbenzoate ≥97%
Methyl 4-methoxy-2,3-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203833
CAS Number: 5628-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR